Octyl isocyanate ≥97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Gevaar

L18746.06
L18746.06EA 73.9 EUR
L18746.06
Octyl isocyanate ≥97%
Octyl isocyanate
Formule: C₉H₁₇NO
Kookpunt: 200…204 °C
Dichtheid: 0,880
Vlampunt: 71 °C (159 °F)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00013887
CAS nummer: 3158-26-7
EINECS: 221-598-7
UN: 2206
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR