L-Proline methyl ester hydrochloride ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L19290.03 L19290.06
L19290.03EA 29.3 EUR
L19290.03 L19290.06
L-Proline methyl ester hydrochloride ≥98%
L-Proline methyl ester hydrochloride

Purity: 98+% CAS No.: 2133-40-6 Molecular Formula: C6H11NO2.HCl Formula Weight: 165.62 Storage Temperature: 2°C to 8°C Melting Point: 71-78° Beilstein Reference: 3596045 Optical Rotation: -33° (c=1 in water) MDL No.: MFCD00012708

Formule: C₆H₁₁NO₂·HCl
Smeltpunt: 71…78 °C
MDL Nummer: MFCD00012708
CAS nummer: 2133-40-6
EINECS: 218-363-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR