2-Fluoro-4-nitrobenzoic acid ≥98%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L19770.03 L19770.06
L19770.03EA 73.5 EUR
L19770.03 L19770.06
2-Fluoro-4-nitrobenzoic acid ≥98%
2-Fluoro-4-nitrobenzoic acid

Purity: 98% CAS No.: 403-24-7 Molecular Formula: C7H4FNO4 Formula Weight: 185.11 Melting Point: 172-175° Beilstein Reference: 2582091

Formule: C₇H₄FNO₄
MW: 185.11 g/mol
Smeltpunt: 172…175 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00275565
CAS nummer: 403-24-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR