4-Iodobenzylamine hydrochloride 97%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L19967.03 L19967.06
L19967.03EA 80 EUR
L19967.03 L19967.06
4-Iodobenzylamine hydrochloride 97%
4-Iodobenzylamine hydrochloride

Purity: 97% CAS No.: 59528-27-7 Molecular Formula: C7H8IN.HCl Formula Weight: 269.51 Melting Point: 275-280° dec. Beilstein Reference: 3691299

Formule: C₇H₉ClIN
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD04039324
CAS nummer: 59528-27-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR