2-Methoxy-5-pyridineboronic acid 95%

Leverancier: Thermo Fisher Scientific
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Waarschuwing

L20087.03 L20087.06
L20087.03EA 34.9 EUR
L20087.03 L20087.06
2-Methoxy-5-pyridineboronic acid 95%
2-Methoxy-5-pyridineboronic acid

Purity: 95% CAS No.: 163105-89-3 Molecular Formula: C6H8BNO3 Formula Weight: 152.95 Melting Point: 132-133° Beilstein Reference: 9119950

Formule: C₆H₈BNO₃
Smeltpunt: 132…133 °C
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD02093044
CAS nummer: 163105-89-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR