Lucifer Yellow VS dilithium salt ~85%, Sigma-Aldrich®

Leverancier: SIGMA ALDRICH MICROSCOPY
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: 4-Amino-N-(3-[vinylsulphonyl]phenyl)naphthalimide-3,6-disulphonate dilithium salt, Dilithium 4-amino-N-[3-(vinylsulphonyl)phenyl]naphthalimide-3,6-disulphonate

M-Clarity™ quality level = MQ100

L3510-100MG L3510-25MG
L3510-100MGEA 505 EUR
L3510-100MG L3510-25MG
Lucifer Yellow VS dilithium salt ~85%, Sigma-Aldrich®
Lucifer Yellow VS dilithium salt
Formule: C₂H₁₂Li₂N₂OS₃
MW: 550.39 g/mol
Bewaar temperatuur: Room Temperature
MDL Nummer: MFCD00009775
CAS nummer: 71231-14-6
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b65df379ff69fd5ab04af7580bc34cea471155b5&vid=9fd0c5e2ae559805b854fd29376937dfcedf6305&prid=web&clid=SAPDAM

Order Now

Specification Test Results

Appearance (Colour) Yellow to Brown
Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Solubility (Colour) Light Yellow to Brown
Solubility (Turbidity) Clear to Slightly Hazy
Purity (HPLC) ≥80.00 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR