Bi-directional/Linear shakers, Varioshake series
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
L003058 L003062
LAUDL003058EA 1520 EUR
LAUDL003058 LAUDL003062
Bi-directional/Linear shakers, Varioshake series
Schudapparaten Schudapparaten met heen-en-weergaande beweging
Shakers with bi-directional/linear motion, heavy duty, low noise, in a compact design.
  • Shaking frequency display and setting digitally via LC display, in steps of 1,0 min⁻¹, gentle start-up
  • The setpoint and actual values ​​as well as the shaking frequency are displayed on two LC displays
  • User-friendly microprocessor technology
  • Membrane keyboard with distinctive symbols
  • Timer adjustable from 1 min up to 99:59 hours or continuous operation
  • Optional RS232 interface on request
  • Large electrolytically galvanised, powder-coated sheet steel shaking platform
  • Four