Heat Transfern Liquid, Ultra 301
Leverancier: LAUDA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
LZB353
LAUDLZB353EA 0 EUR
LAUDLZB353
Heat Transfern Liquid, Ultra 301
Vloeistoffen voor circulatiebaden
Heat transfer liquid for temperatures up to over 300 °C.
  • Silicone-free liquid, ideal for surface-sensitive applications
  • The safety data sheet must be observed for uses in bath thermostats