Decontamination spray, Diphoterine
Leverancier: LABORATOIRES ESCULAPE
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
8205211
LBES8205211EA 235 EUR
LBES8205211
Decontamination spray, Diphoterine
Eerste-hulpbenodigdheden Douches
These Diphoterine® solution aerosol sprays are used to wash small chemical splashes. A 100 ml aerosol spray is sufficient to wash the body surface area equal to that of one hand. A 200 ml aerosol spray is sufficient to wash the surface area equal to an arm or a face.
  • To be used within one minute after the splash
  • Must be stored in any area where the risk due to small chemical projections exists