Joint clamps for conical ground joints, Safety Clip
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01103845 01103829 01103814 01103812 01103819 01103824 01103834
LENZ01103845EA 4.1 EUR
LENZ01103845 LENZ01103829 LENZ01103814 LENZ01103812 LENZ01103819 LENZ01103824 LENZ01103834
Joint clamps for conical ground joints, Safety Clip
Klemmen NS-verbindingsklemmen
Clamps made of POM, for firm and secure fitting of conical ground joints.
  • Will not scratch glass surfaces
  • Suitable for use with sleeves with grip collar
  • Outstanding chemical resistance
  • Operating temperature up to 150 °C