Adapters, glass, cone/screwthread
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
01301819 01301419 01301814 01302519 01302524 01303229 01301414 01301424 01301429 01301829 01302529
201-2116EA 22.7 EUR
201-2116 LENZ01301419 LENZ01301814 LENZ01302519 LENZ01302524 LENZ01303229 LENZ01301414 LENZ01301424 LENZ01301429 LENZ01301829 LENZ01302529
Adapters, glass, cone/screwthread
Adapters Adapters met NS-verbindingen
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Screw thread adapters, with screw-cap with aperture and silicone sealing ring with PTFE washers.