Afsluitkranen, driewegversie, Czako
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
02631804
LENZ02631804EA 107 EUR
LENZ02631804
Afsluitkranen, driewegversie, Czako
Afsluitkranen
Stopcocks, made of DURAN® borosilicate glass.
  • Good temperature resistance
  • Good chemical resistance
With PTFE plug and screwthread retaining device.
Certificaten: Leakage rate according to DIN 12540-1.