Destillatiekoppelingen met spatbol
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05043002 05043003 05043004 05043001
LENZ05043002EA 114 EUR
LENZ05043002 LENZ05043003 271-3102 LENZ05043001
Destillatiekoppelingen met spatbol
Onderdelen voor destillatie Destillatiebruggen
DURAN®, borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
With two vertical NS cones and splash head.