Condensators, Davies
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05215303 05215304 05215403 05215404
LENZ05215303EA 119 EUR
LENZ05215303 LENZ05215304 LENZ05215403 LENZ05215404
Condensators, Davies
Condensors
DURAN® borosilicate glass 3.3.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
For condensing vapours. With double sized coiling area. Water connection with two glass olives.