Adapters, levering, recht, met afvoertip
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
05266003 05266002 05266001 05266004
LENZ05266003EA 25.1 EUR
LENZ05266003 LENZ05266002 LENZ05266001 LENZ05266004
Adapters, levering, recht, met afvoertip
Onderdelen voor destillatie Destillatieadapters
DURAN®, borosilicate glass.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Delivery adapters, straight, with NS socket and NS cone, with drain tip.