Flat flange lids, centre neck
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
06100204 06100104 06100201 06100206 06100404 06100406 06100504 06100506
LENZ06100204EA 179 EUR
LENZ06100204 LENZ06100104 LENZ06100201 LENZ06100206 LENZ06100404 LENZ06100406 LENZ06100504 LENZ06100506
Flat flange lids, centre neck
Deksels Deksels voor reactievaten
DURAN®, Borosilicate glass.
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion giving relatively high resistance to temperature changes
Flange lids for reaction vessels, with laboratory flange LF.