Erlenmeyers, smalle hals, met rolrand
Leverancier: Lenz Laborglas GmbH & CO.KG
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
07020022 07020028 07020037 07020046 07020049 07020052 07020058 07020070 07020073 07020076 07020082
LENZ07020022EA 112 EUR
LENZ07020022 LENZ07020028 LENZ07020037 LENZ07020046 LENZ07020049 LENZ07020052 LENZ07020058 LENZ07020070 LENZ07020073 LENZ07020076 LENZ07020082
Erlenmeyers, smalle hals, met rolrand
Kolven Erlenmeyers
DURAN®, borosilicate glass 3.3, clear.
  • Very good chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Volume graduations and labelling area.
Order Now
Specificaties
Environmentally Preferable