Biological indicators, stainless steel coupons, OXI CONTROL
Leverancier: LIOFILCHEM
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
91082 91097 91047 91048 91079 91080 91081 91083 91085 91086 91088 91089 91091 91092 91096 91098 91180 91182
91082.EA 353 EUR
91082. LIOF91097 LIOF91047 LIOF91048 LIOF91079 LIOF91080 LIOF91081 LIOF91083 LIOF91085 LIOF91086 LIOF91088 LIOF91089 LIOF91091 LIOF91092 LIOF91096 LIOF91098 LIOF91180 LIOF91182
Biological indicators, stainless steel coupons, OXI CONTROL
Sterilisatie-indicatoren
Biological indicators for validating the sterilisation using VHP in isolators.