FlashGel® camera
Leverancier: LONZA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
57040 57062
730-1346EA 1330 EUR
730-1346 LONZ57062
FlashGel® camera
Fotodocumentatiesystemen
The FlashGel® camera captures data from the FlashGel® system, allowing complete separation, recovery and documentation safely, at the bench, in minutes. This simple digital camera in an enclosed hood connects directly to a laptop, PC or PDA via USB.
  • Complete gel run and image capture in five minutes
  • Photograph gels at the bench, without UV light
  • Sharp, clear, high-resolution images
  • Simple user interface
  • Optimised exposure for FlashGel® cassettes
Hood (W×L×H): 100×110×160 mm
Image file type: .JPG, .TIFF, .PDF