Verification standard kit, MultiDirect
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
215650 215670
LOVI215650EA 199.9 EUR
LOVI215650 LOVI215670
Verification standard kit, MultiDirect
Standaarden Standaarden voor kleurreferentie en spectroscopie
Verification accuracy of photometers and validity of results.
  • Check the performance of photometers over the full measurement range
  • Results can be checked against tolerances provided on the certificate of conformity
Leveringsinformatie: Contains zero solution in sealed photometer vial, 6× verification standard solution in sealed photometer vial, light shield cap, certificate of conformity, instruction sheet.