pH indicator, bromophenol blue tablets
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
511621 511620 221030
162623AEA 78.6 EUR
162623A LOVI511620 LOVI221030
pH indicator, bromophenol blue tablets
Milieutestkits
Indicator for pH range 2,8 to 4,4.
Can be used for measuring pH in conjunction with the Comparator 2000+ system and pH disc 2/1B