Silt density index kit
Leverancier: Lovibond
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
56K001901
LOVI56K001901EA 697.4 EUR
LOVI56K001901
Silt density index kit
Milieutestkits
The Silt Density Index (SDI) is a measure of the solids or particulate content of water and is of specific importance when commissioning reverse osmosis plant.
  • SDI calculations in just 15 minutes
  • Ideally suited for reverse osmosis plants
  • Enables calculation of membrane fouling time, backwash frequency and membrane lifespan
The kit utilises a membrane/flow meter system to enable SDI calculations in just 15 minutes. Sufficient membranes included for 100 tests.
Order Now
Specificaties
Environmentally Preferable