Ferroin (1,10-phenanthrolineferrous sulphate complex) solution_SC N1 Indicator

Leverancier: Lovibond
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

Synoniemen: Tris(1,10-phenanthroline)iron (II) sulphate, Tris(1,10-phenanthroline-N1,N10)iron sulphate

56L017165
LOVI56L017165EA 24.9 EUR
LOVI56L017165
Ferroin (1,10-phenanthrolineferrous sulphate complex) solution_SC N1 Indicator
Ferroin (1,10-phenanthrolineferrous sulphate complex)
Formule: C₃₆H₂₄FeN₆SO₄
MW: 692.54 g/mol
Dichtheid: 0.9999
MDL Nummer: MFCD00036428
CAS nummer: 14634-91-4

Order Now

Specificaties

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR