Graduated pipettes, disposable
Leverancier: LP ITALIANA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
160118
LPIT160118EA 332 EUR
LPIT160118
Graduated pipettes, disposable
Pipetten Gegradueerde pipetten
Disposable graduated plastic pipettes with open end.
  • Sterile (Ionising Radiation)