Test tube racks, acetal resin
Leverancier: LP ITALIANA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
189082 187082 187083 187084 187086 188083 188084 188085 188086 189083 189084 189085 189086 188081
LPIT189082EA 286 EUR
LPIT189082 LPIT187082 LPIT187083 LPIT187084 LPIT187086 LPIT188083 LPIT188084 LPIT188085 LPIT188086 LPIT189083 LPIT189084 LPIT189085 LPIT189086 LPIT188081
Test tube racks, acetal resin
Rekken Tube Racks
Made of Acetal resin.
  • Good chemical resistance
  • Temperature up to 121 °C.
Square hole racks.
Order Now
Specificaties
Environmentally Preferable