Bottle Carriers
Leverancier: LP ITALIANA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
291187
LPIT291187EA 24.5 EUR
LPIT291187
Bottle Carriers
Dragers Flessendragers
Bottle carrier and container for bottles or tools, made of red PP.
  • Good temperature resistance
  • Good chemical resistance
  • Sturdy
Order Now
Specificaties
Environmentally Preferable