Draining racks
Leverancier: LP ITALIANA
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
291195
LPIT291195EA 101 EUR
LPIT291195
Draining racks
Rekken Afdruiprekken
Drying rack, made of PP, with 81 places that can be fully disassembled.
  • Excellent chemical resistance
  • Excellent temperature resistance
  • With carrying handle