Buisdop, LABOCAP, zonder handvat
Leverancier: LUDI
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
184110521 184110522 184110523 184110631 184110632 184110633 184110741 184110742 184110743 184110851 184111411 184111832 184111941 184112051 184112161 184111831 184112162 184110852 184112052 184112053 184110635 184111943 184110744 184110853
391-5901EA 16.1 EUR
391-5901 391-5903 391-5902 391-5907 391-5909 391-5908 391-5910 391-5912 391-5911 391-5913 LUDI184111411 391-5906 LUDI184111941 LUDI184112051 LUDI184112161 LUDI184111831 LUDI184112162 LUDI184110852 LUDI184112052 LUDI184112053 LUDI184110635 LUDI184111943 LUDI184110744 LUDI184110853
Buisdop, LABOCAP, zonder handvat
Sluitingen Reageerbuissluitingen
Made of anodised aluminum with stainless steel spring clip.
  • Springs clamp against the outer wall of the vessel
  • Anodised and coloured
  • Autoclavable
Without grip. Cap for sterile but not hermetic closing of test tubes and other vessels.