Ammonium sulphate ≥99.5%, Proteomics Grade
Leverancier: VWR Chemicals
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
Waarschuwing
Synoniemen: Sulphuric acid diammonium salt, Diammonium sulphate, Mascagnite
M105-1KGEA 110 EUR
M105-1KG
Ammonium sulphate ≥99.5%, Proteomics Grade
Ammonium sulphate
Formule: (NH₄)₂SO₄
Smeltpunt: 336…339 °C
Dichtheid: 1,78 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00003391
CAS nummer: 7783-20-2
EINECS: 231-984-1
Bereik: 01-2119455044-46
Merck Index: 13,00559
Order Now
Specification Test Results
Arsenic <0.0001%
Chloride <0.0005%
DNase none detected
Heavy Metals (as Pb) <0.0002%
Insolubles <0.005%
Iron <0.0005%
Magnesium Test Pass
pH (5%, water) @25 °C 5.0 – 6.0
Phosphate <0.0002%
Protease none
Purity (Dry Basis) >99.5%
Residue on Ignition <0.005%
RNase none detected
Selenium ≤0.0005%