Mag-Bind® Total RNA 96 kit
Leverancier: OMEGA BIO-TEK
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
M6731-01 M6731-00
M6731-01EA 1320 EUR
M6731-01 M6731-00
Mag-Bind® Total RNA 96 kit
Reagentia voor nucleïnezuren Nucleic Acid Purification Kits and Reagents
Isolate total cellular RNA and viral RNA from a wide variety of cells and tissues.
  • Procedure can be fully automated with most robotic workstations
The Mag-Bind® Total RNA 96 Kit provides a novel technology for total RNA isolation. This kit allows the rapid and reliable isolation of high-quality total cellular RNA and viral RNA from a wide variety of cells and tissues. Unlike column-based systems, the binding of nucleic acids to magnetic particles occurs in solution resulting in increased binding kinetics and binding efficiency. Particles are also completely re-suspended during the wash steps of the purification protocol, which improves the removal of contaminants and increases nucleic acid purity. Mag-Bind® Total RNA 96 Kit procedure can be fully automated with most robotic workstations.