Magnesium chloride hexahydrate 99.0-102.0% ACS, Fluka™

Leverancier: Honeywell Chemicals
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

M9272-100G M9272-1KG M9272-2.5KG M9272-4X2.5KG M9272-500G M9272-6X500G
M9272-100GEA 33.7 EUR
M9272-100G M9272-1KG M9272-2.5KG M9272-4X2.5KG M9272-500G M9272-6X500G
Magnesium chloride hexahydrate 99.0-102.0% ACS, Fluka™
Magnesium chloride hexahydrate
Formule: MgCl₂·6H₂O
Kookpunt: 1412 °C (1013 hPa)
Smeltpunt: 117 °C (Elimination of water of crystallisation)
Dichtheid: 1,57 g/cm³ (20 °C)
Bewaar temperatuur: Kamertemperatuur
MDL Nummer: MFCD00149781
CAS nummer: 7791-18-6
EINECS: 232-094-6
Merck Index: 13,05684

Order Now

Specification Test Results

Assay 99.0 - 102.0 %
Water insoluble matter Max. 0.005 %
Barium (Ba) Max. 0.005 %
Calcium (Ca) Max. 0.01 %
Iron (Fe) Max. 5 ppm
Potassium (K) Max. 0.005 %
Manganese (Mn) Max. 5 ppm
Sodium (Na) Max. 0.005 %
Strontium (Sr) Max. 0.005 %
Heavy metals (as Pb) Max. 5 ppm
Nitrate (NO₃) Max. 0.001 %
Phosphate (PO₄) Max. 5 ppm
Sulfate (SO₄) Max. 0.002 %
Ammonium (NH₄) Max. 0.002 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR