Qualitative filter papers, technical, smooth, grade MN 713
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4007130018 400713002 4007130015 400713004 4007130085 400713018 400713007 4007130022 4007130035
MANA4007130018EA 5.8 EUR
MANA4007130018 MANA400713002 MANA4007130015 MANA400713004 MANA4007130085 MANA400713018 MANA400713007 MANA4007130022 MANA4007130035
Qualitative filter papers, technical, smooth, grade MN 713
Filters Filterpapier Kwalitatief filterpapier
Smooth filter papers particularly suited for technical applications such as industrial filtrations.
Medium fast filtration, suitable for many applications.