Glass wool
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
706201 718587 718002
MANA706201EA 109 EUR
MANA706201 MANA718587 MANA718002
Glass wool
Glaswol en katoen
This glass wool can be used for GC liners, packed GC columns etc.