HPLC columns, NUCLEOSIL® C8
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
720735.46 720990.46 721194.30 720001.40 720001.46 720013.40 720013.46 720017.46 720022.40 720022.46 720050.40 720050.46 720052.40 720052.46 720062.46 720071.40 720071.46 720214.46 720165.46 721096.30 721061.30 721093.30 721095.30
MANA720735.46EA 418 EUR
MANA720735.46 MANA720990.46 MANA721194.30 MANA720001.40 MANA720001.46 MANA720013.40 MANA720013.46 MANA720017.46 MANA720022.40 MANA720022.46 MANA720050.40 MANA720050.46 MANA720052.40 MANA720052.46 MANA720062.46 MANA720071.40 MANA720071.46 MANA720214.46 554-5800 554-5805 MANA721061.30 MANA721093.30 MANA721095.30
HPLC columns, NUCLEOSIL® C8
Kolommen voor chromatografie
NUCLEOSIL® C8 and C8 ec are octyl modified silica phases for HPLC.
  • Robust spherical silica gel
  • Weakly hydrophobic, polar, endcapped or non-endcapped phases for RP chromatography
  • Separation mechanism based on hydrophobic (van der Waals) and slight or noticeable residual silanol interactions
EC guard columns are used for analytical EC columns with the Column Protection System guard column holder (554-5851).