SPE columns, CHROMABOND® Alox
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
730139 730139G 730250 730446 730453 730467 730017 730020 731844 731845 732091 738252.100M
MANA730139EA 186 EUR
MANA730139 MANA730139G MANA730250 MANA730446 MANA730453 MANA730467 MANA730017 MANA730020 MANA731844 MANA731845 MANA732091 MANA738252.100M
SPE columns, CHROMABOND® Alox
Kolommen voor chromatografie
CHROMABOND® Alox is a high pure aluminium oxide for SPE.
  • Available as acidic, neutral or basic material
  • Polar phase
  • Alternative selectivity to SiOH
Recommended for polar substances like e.g. lipophylic dyes.