SPE columns, CHROMABOND® HR-XCW
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
730743 730741 730731 730731P45 730733 730733P45 730735 730735P45 730737 730739 730739P45 731774 731775 731776
MANA730743EA 275 EUR
MANA730743 MANA730741 MANA730731 MANA730731P45 MANA730733 MANA730733P45 MANA730735 MANA730735P45 705-1114 705-1113 MANA730739P45 MANA731774 MANA731775 MANA731776
SPE columns, CHROMABOND® HR-XCW
Kolommen voor chromatografie
CHROMABOND® HR-XCW is a weak carboxylic acid modified polymeric cation exchanger for SPE.
  • Base material: Spherical PS/DVB copolymer with a large specific surface and a high pH stability of 1 to 14
  • Cation exchange capacity >0,7 meq/g
  • Outstanding recovery rates especially for basic analytes
The weak polymeric cation exchanger is recommended for basic compounds (quaternary amines, strong bases with pKa >10) from strongly matrix-contaminated samples.