Electronic crimping tools
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
735511 735520 735611 735620 735513 735532 735533 735613
MANA735511EA 1100 EUR
MANA735511 MANA735520 MANA735611 MANA735620 MANA735513 MANA735532 MANA735533 MANA735613
Electronic crimping tools
Capsuleer- en decapsuleertangen
These battery-powered electronic crimping tools are suitable for aluminium crimp and Flip Top / Flip Off caps (not suitable for 20 mm magnetic / bi-metal crimp caps).
  • Mobile tools for consistent and reproducible crimping results
  • Crimping pressure adjustable by the control unit on top of the tool
  • Long lasting lithium ion cell batteries (full battery charge for several hundred vials, life time of battery >1500 charges)
Certificaten: CE certificate of conformity along with one year warranty.