HPLC columns, NUCLEODUR® Phenyl-Hexyl
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
760589.46 760566.46 760586.20 760561.46 760566.40 760561.40 760563.20 760563.30 760563.40 760563.46 760565.20 760566.20 760566.30 760568.20 760573.20 760573.46 760575.46 760576.20 760576.30 760576.46 760577.20 760577.30 760577.40 760577.46 760578.20 760578.30 760578.40 760578.46 760579.30 760579.40 760579.46 760583.20 760583.30 760583.46 760586.30 760586.46 760587.30 760587.40 760587.46 760588.20 760588.30 760588.40 760588.46 760589.30 760589.40 761985.20 761985.30 761986.20 761986.30 761987.20 761987.30 760585.46 760587.20 760589.20 760561.20 760576.40 760579.20 760573.30 760583.40 760561.30 760573.40 760586.40
MANA760589.46EA 814 EUR
MANA760589.46 MANA760566.46 MANA760586.20 MANA760561.46 MANA760566.40 MANA760561.40 MANA760563.20 MANA760563.30 MANA760563.40 MANA760563.46 MANA760565.20 MANA760566.20 MANA760566.30 MANA760568.20 MANA760573.20 MANA760573.46 MANA760575.46 MANA760576.20 MANA760576.30 MANA760576.46 MANA760577.20 MANA760577.30 MANA760577.40 MANA760577.46 MANA760578.20 MANA760578.30 MANA760578.40 MANA760578.46 MANA760579.30 MANA760579.40 MANA760579.46 MANA760583.20 MANA760583.30 MANA760583.46 MANA760586.30 MANA760586.46 MANA760587.30 MANA760587.40 MANA760587.46 MANA760588.20 MANA760588.30 MANA760588.40 MANA760588.46 MANA760589.30 MANA760589.40 MANA761985.20 MANA761985.30 MANA761986.20 MANA761986.30 MANA761987.20 MANA761987.30 MANA760585.46 MANA760587.20 MANA760589.20 MANA760561.20 MANA760576.40 MANA760579.20 MANA760573.30 MANA760583.40 MANA760561.30 MANA760573.40 MANA760586.40
HPLC columns, NUCLEODUR® Phenyl-Hexyl
Kolommen voor chromatografie
NUCLEODUR® Phenyl-Hexyl is a phenyl-hexyl modified silica phase with multi-endcapping for HPLC.
  • High purity spherical silica gel
  • Hydrophobic RP phase with aromatic selectivity
  • Separation mechanism based on hydrophobic and π-π interactions
With the separation principle based on two retention mechanisms NUCLEODUR® Phenyl-Hexyl is an alternative to classical C18 modifications. Thus, this phase is well suited for aromatic and unsaturated compounds like aromatic acids, steroids, and flavonoids - also for LC-MS detection.
EC guard columns are used forMore Product Information