HPLC columns, NUCLEODUR® C8 ec
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
760700.20 760700.30 760700.40 760700.46 760701.20 760701.30 760701.40 760701.46 760702.46 760703.20 760703.30 760703.40 760703.46 760060.20 760060.30 760060.40 760060.46 760061.46 760062.20 760062.30 760062.40 760062.46 760063.20 760063.30 760063.40 760063.46 760704.20 760704.30 760704.40 760704.46 760706.46 761936.20 761936.30 761937.20 761937.30 762061.100 762062.100 762062.210 762072.100 762092.160 762092.80 760059.20 760059.30 760059.40 760059.46 760064.46
MANA760700.20EA 375 EUR
MANA760700.20 MANA760700.30 MANA760700.40 MANA760700.46 MANA760701.20 MANA760701.30 MANA760701.40 MANA760701.46 MANA760702.46 MANA760703.20 MANA760703.30 MANA760703.40 MANA760703.46 MANA760060.20 MANA760060.30 MANA760060.40 MANA760060.46 MANA760061.46 MANA760062.20 MANA760062.30 MANA760062.40 MANA760062.46 MANA760063.20 MANA760063.30 MANA760063.40 MANA760063.46 MANA760704.20 MANA760704.30 MANA760704.40 MANA760704.46 MANA760706.46 MANA761936.20 MANA761936.30 MANA761937.20 MANA761937.30 MANA762061.100 MANA762062.100 MANA762062.210 MANA762072.100 MANA762092.160 MANA762092.80 MANA760059.20 MANA760059.30 MANA760059.40 MANA760059.46 MANA760064.46
HPLC columns, NUCLEODUR® C8 ec
Kolommen voor chromatografie
NUCLEODUR® C8 ec is a medium density monomeric octyl modified silica phase for HPLC.
  • High purity spherical silica gel
  • Weakly hydrophobic, endcapped phase with weakly polar selectivity for RP chromatography
  • Separation mechanism based on slight hydrophobic interactions
The RP phase is ideal for daily routine analysis with recommended application such as the separations of drugs, pesticides, fat-soluble vitamins, aldehydes and ketones as well as phenolic compounds.
EC guard columns are used for analytical EC columns with the Column Protection SystemMore Product Information
Order Now
Specificaties
Environmentally Preferable
Koolstofgehalte 10,5%
Endcapping Ja
pH-bereik 1 - 9
USP code L7