TLC introductory kit, Micro Set A
Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
814002 814003 814000 814001 814011 814012 814013
MANA814002EA 29.4 EUR
MANA814002 MANA814003 552-0089 MANA814001 MANA814011 MANA814012 MANA814013
TLC introductory kit, Micro Set A
Educatief materiaal Educatief materiaal over chemie Leeractiviteiten voor chromatografie
An ideal introductory kit for the science education sector. The kit contains all the the chemicals, plates and accessories for students to perform separations of dyes including: anthaquinone dye mixtures, lipophilic dye mixtures and food dye mixtures. The TLC plates are polyester sheet based with either silica, cellulose or alumina as coating.
Leveringsinformatie: Kit includes 3 developing tanks, 50 glass capillaries (1 µl), 1 spotting guide, 1×10 ml measuring cylinder, 50 sheets 40×80 mm POLYGRAM SIL G/UV₂₅₄, 50 sheets 40×80 mm ALOX N/UV₂₅₄ and 50 sheets 40×80 mm of CEL 300, 3 dyeMore Product Information