NANOCOLOR® Photometric Tube Test Kits for Nitrate

Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

985064 985066 985065
MANA985064EA 107 EUR
MANA985064 MANA985066 MANA985065
NANOCOLOR® Photometric Tube Test Kits for Nitrate
Milieutestkits

Photometric test kits for the determination of nitrate in water samples.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting


The test kits are evaluated in 16 mm (OD) tubes. They are preprogrammend on NANOCOLOR® photometers and also compatible with METTLER TOLEDO Easy VIS and Easy UV. Programming data for other spectrophotometers are available.


Sea water analysis: No
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II


Leveringsinformatie: Rugged box with 20 test tubes.

Order Now

Specificaties

Model MANA985064 MANA985066 MANA985065
Environmentally Preferable
Platform NANOCOLOR® tube tests NANOCOLOR® tube tests NANOCOLOR® tube tests
Opslagtemperatuur 15…25 °C 15…25 °C 15…25 °C
Houdbaarheid (in maanden) 24 24 24
Gewicht 461 g (including packaging) 481 g (including packaging) 464 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR