NANOCOLOR® Photometric Tube Test Kit for Starch

Leverancier: MACHEREY-NAGEL
Ratings: (No Reviews)

Total Ratings: 0
Avg. Ratings: out of 5

985085
MANA985085EA 88.5 EUR
MANA985085
NANOCOLOR® Photometric Tube Test Kit for Starch
Milieutestkits

Photometric test kit for the determination of starch in water samples.


  • Measurement directly in the test tube, saving time and reducing exposure to chemical reagents
  • Bar code cuvette identification for easy test call up in NANOCOLOR® spectrophotometers
  • Precisely pre-dosed reagents with constant high quality from batch to batch
  • Coloured pictogram instructions and easy handling of the tubes
  • 16 mm (OD) tubes for easy pipetting


The test kit is evaluated in 16 mm (OD) tubes. It is preprogrammend on NANOCOLOR® photometers. Programming data for other spectrophotometers are available.


Sea water analysis: Yes
Hazardous material: Yes
Remark: Measuring range on NANOCOLOR VIS II


Leveringsinformatie: Rugged box with 19 test tubes.

Order Now

Specificaties

Model MANA985085
Environmentally Preferable
Platform NANOCOLOR® tube tests
Opslagtemperatuur 15…25 °C
Houdbaarheid (in maanden) 12
Gewicht 383 g (including packaging)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Meer About VWR