Adhesion slides, HistoBond®
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
0810701
MARI0810701EA 793 EUR
MARI0810701
Adhesion slides, HistoBond®
Object- en dekglaasjes Adhesie-objectglaasjes
HistoBond slides combine frosted ends with printed in various colours.
  • Soda lime glass of hydrolytic class 3
  • Different colours for colour coding of methods of analysis
  • Thin-coated marking area facilitates the processing on automated systems
Certificaten: IVD directive 98/79/EC