Capillary tubes
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
2951300 2951303
MARI2951300EA 0 EUR
MARI2951300 MARI2951303
Capillary tubes
Diagnostische systemen Clinical Sample Collection/Preparation Blood Sample Collection/Preparation
Glass, clear.
  • Na-heparinized (coated)
Capillaries, Na-heparinized (coated)
For blood gas analysis.