Petri dishes, soda lime glass
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3400300 3401000 3401100 3401600 3401300
631-0405EA 278 EUR
631-0405 631-0406 MARI3401100 MARI3401600 MARI3401300
Petri dishes, soda lime glass
Cultuurschalen Petrischalen
Soda-lime glass.
  • Free of cadmium and lead
  • Base and cover lid both very flat, reducing optical distortion and facilitating the uniform spreading of nutrient media and liquids
  • Sterilisable up to 121 °C
  • Multiple use