Test tubes with thread
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
3704679
MARI3704679EA 0 EUR
MARI3704679
Test tubes with thread
Buisjes Reageerbuisjes
Soda-lime glass.
  • Good chemical and physical properties
  • Without screw cap
Culture medium tubes with thread for aluminium screw caps are suitable for transporting and growing of cultures.
Bestelinformatie: Screw caps must be ordered seperately.