Glass beads
Leverancier: MARIENFELD
Ratings:
Avg. Ratings: 4.0 (1 Review)
Total Ratings: 1
Avg. Ratings: out of 5
4901003 4901006 4901001 4901002 4901004 4901005
MARI4901003EA 16.9 EUR
MARI4901003 MARI4901006 MARI4901001 MARI4901002 MARI4901004 MARI4901005
Glass beads
Parels Glasparels
Made of soda lime glass.
  • Solid
  • Polished
Glass beads are used for mixing or milling. The smaller the beads and the higher their quantity the more intense is the mixing or milling.
Verpakking: In poly bag of 500 g or 1000 g each.