Slide mailers
Leverancier: MARIENFELD
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
5600065 5610067
MARI5600065EA 57.2 EUR
MARI5600065 MARI5610067
Slide mailers
Verzenddoosjes voor objectglaasjes
Slide mailers made of polyethylene for archiving and staining of microscope slides.
  • Secure closure and space for ²/₅ slides
  • Break resistant rails for easy accommodation
  • Transport of slides without contacting each other