Veiligheidsmes, SECUNORM PROFI25 MDP
Leverancier: MARTOR ARGENTAX
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
120700.02
MARO120700.02EA 36.7 EUR
MARO120700.02
Veiligheidsmes, SECUNORM PROFI25 MDP
Messen
Trapeziummesje, aluminium handgreep.
  • With spring loaded blade retraction, designed to respond to any uncontrolled move if and when the blade slider is not in forward position
  • Can be used by left and right-handed users
  • Easy blade change: No extra tools needed
  • Stainless 'food contact approved' blades
This safety cutter with aluminium body with metal-detectable blue plastic parts and stainless 'food contact approved' blades is ideal for slitting cardboard and adhesive tape.
TÜV tested
Certificaten: TÜV-goedgekeurd.