Cap mats and strips
Leverancier: THERMO MATRIX TECHNOLOGIES
Ratings: (No Reviews)
Total Ratings: 0
Avg. Ratings: out of 5
4411 4412 4413 4414 4415 4416 4421 4422 4410 4430 4431 4432
MATR4411EA 51.5 EUR
MATR4411 MATR4412 MATR4413 MATR4414 MATR4415 MATR4416 MATR4421 MATR4422 MATR4410 MATR4430 MATR4431 MATR4432
Cap mats and strips
Afdichtingen Afdichtingen van microtiterplaten
Matrix CapMats and CapStrips are engineered to maintain a strong seal at all times on tubes, microplates, and deepwell blocks. CapMats effectively seal down to –80 °C, while cap strips give the flexibility to seal individual caps or rows.
  • Low temperature compatible so seals remain strong even under extreme conditions
  • Securely seals plates, blocks and tubes ensures sample integrity no matter what the vessel
  • EVA material construction provides a chemically resistant seal
  • Alphanumeric designation on mats simplifies well identification
Cap mats are theMore Product Information